Assay Details
| Gene Symbol: rnf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017071.2 | NP_001017071.1 | 1 - 2 | 112 | 71 | |
| GenBank mRNA | CR761547.2 | - | 1 - 2 | 112 | 71 | |
More Information
Set Membership: |
|

rnf5
Genomic Map