Assay Details
| Gene Symbol: mrpl51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017085.2 | NP_001017085.1 | 3 - 4 | 237 | 88 | |
| XM_012959079.2 | XP_012814533.1 | 3 - 4 | 280 | 88 | ||
| GenBank mRNA | CR761437.2 | - | 3 - 4 | 237 | 88 | |
More Information
Set Membership: |
|

mrpl51
Genomic Map