Assay Details
| Gene Symbol: rplp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017204.2 | NP_001017204.1 | 2 - 3 | 217 | 86 | |
| GenBank mRNA | BC118872.1 | - | 2 - 3 | 211 | 86 | |
| CR760503.2 | - | 2 - 3 | 217 | 86 | ||
More Information
Set Membership: |
|

rplp1
Genomic Map