Assay Details
| Gene Symbol: supt16h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017230.2 | NP_001017230.1 | 4 - 5 | 604 | 77 | |
| GenBank mRNA | CR760255.2 | - | 4 - 5 | 604 | 77 | |
More Information
Set Membership: |
|

supt16h
Genomic Map