Assay Details
| Gene Symbol: rdh14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017231.2 | NP_001017231.1 | 1 - 2 | 394 | 73 | |
| GenBank mRNA | BC157371.1 | - | 1 - 2 | 318 | 73 | |
| CR760254.2 | - | 1 - 2 | 394 | 73 | ||
More Information
Set Membership: |
|

rdh14
Genomic Map