Assay Details
| Gene Symbol: tmem165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017260.2 | NP_001017260.1 | 2 - 3 | 300 | 61 | |
| GenBank mRNA | BC161030.1 | - | 2 - 3 | 343 | 61 | |
| CR760016.2 | - | 2 - 3 | 300 | 61 | ||
More Information
Set Membership: |
|

tmem165
Genomic Map