Assay Details
| Gene Symbol: gtf2h5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017298.2 | NP_001017298.1 | 2 - 3 | 161 | 86 | |
| GenBank mRNA | CR855603.2 | - | 2 - 3 | 161 | 86 | |
More Information
Set Membership: |
|

gtf2h5
Genomic Map