Assay Details
| Gene Symbol: cdc42se2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030323.2 | NP_001025494.1 | 2 - 3 | 535 | 69 | |
| GenBank mRNA | BC123961.1 | - | 1 - 2 | 219 | 69 | |
| CR762307.2 | - | 2 - 3 | 535 | 69 | ||
More Information
Set Membership: |
|

cdc42se2
Genomic Map