Assay Details
| Gene Symbol: cdc42se2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030333.2 | NP_001025504.1 | 2 - 3 | 296 | 65 | |
| GenBank mRNA | CR761398.2 | - | 2 - 3 | 296 | 65 | |
More Information
Set Membership: |
|

cdc42se2
Genomic Map