Assay Details
| Gene Symbol: psmd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037272.1 | NP_001032349.1 | 1 - 2 | 160 | 65 | |
| GenBank mRNA | BC077668.1 | - | 1 - 2 | 165 | 65 | |
| BC088002.1 | - | 1 - 2 | 160 | 65 | ||
More Information
Set Membership: |
|

psmd7
Genomic Map