Assay Details
| Gene Symbol: guk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039729.1 | NP_001034818.1 | 5 - 6 | 457 | 62 | |
| XM_012964469.2 | XP_012819923.1 | 5 - 6 | 498 | 62 | ||
| GenBank mRNA | BC064272.1 | - | 5 - 6 | 455 | 62 | |
| CR848346.2 | - | 5 - 6 | 457 | 62 | ||
More Information
Set Membership: |
|

guk1
Genomic Map