Assay Details
| Gene Symbol: art5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039735.1 | NP_001034824.1 | 2 - 3 | 847 | 69 | |
| GenBank mRNA | BC088045.1 | - | 3 - 4 | 829 | 69 | |
| BC121650.1 | - | 2 - 3 | 862 | 69 | ||
| CR761378.2 | - | 2 - 3 | 847 | 69 | ||
More Information
Set Membership: |
|

art5
Genomic Map