Assay Details
| Gene Symbol: tnni1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203628.2 | NP_988959.1 | 4 - 5 | 588 | 81 | |
| GenBank mRNA | BC061268.1 | - | 4 - 5 | 599 | 81 | |
| CR760892.2 | - | 4 - 5 | 588 | 81 | ||
More Information
Set Membership: |
|

tnni1
Genomic Map