Assay Details
| Gene Symbol: diablo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016281.3 | NP_001016281.1 | 5 - 6 | 509 | 58 | |
| XM_018093111.1 | XP_017948600.1 | 6 - 7 | 812 | 58 | ||
| GenBank mRNA | BC121212.1 | - | 5 - 6 | 509 | 58 | |
| CR761608.2 | - | 5 - 6 | 528 | 58 | ||
More Information
Set Membership: |
|

diablo
Genomic Map