Assay Details
| Gene Symbol: ppp6c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017110.3 | NP_001017110.1 | 4 - 5 | 414 | 66 | |
| GenBank mRNA | BC123920.1 | - | 4 - 5 | 414 | 66 | |
| CR761231.2 | - | 4 - 5 | 408 | 66 | ||
More Information
Set Membership: |
|

ppp6c
Genomic Map