Assay Details
| Gene Symbol: hes5.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044415.1 | NP_001037880.1 | 269 | 77 | ||
| GenBank mRNA | BC075552.1 | - | 261 | 77 | ||
| CR760038.2 | - | 269 | 77 | |||
| Gene Symbol: LOC101733898 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004916155.3 | XP_004916212.1 | 2 - 3 | 826 | 77 | |
More Information
Set Membership: |
|

hes5.1
Genomic Map