Assay Details
| Gene Symbol: ckb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044461.1 | NP_001037926.1 | 4 - 5 | 546 | 77 | |
| XM_012968533.2 | XP_012823987.1 | 4 - 5 | 583 | 77 | ||
| GenBank mRNA | BC161187.1 | - | 2 - 3 | 1202 | 77 | |
| CR761243.2 | - | 4 - 5 | 546 | 77 | ||
More Information
Set Membership: |
|

ckb
Genomic Map