Assay Details
| Gene Symbol: josd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044464.1 | NP_001037929.1 | 1 - 2 | 164 | 62 | |
| GenBank mRNA | BC155032.1 | - | 1 - 2 | 177 | 62 | |
| BC155035.1 | - | 1 - 2 | 177 | 62 | ||
| BC158992.1 | - | 1 - 2 | 177 | 62 | ||
| CR761270.2 | - | 1 - 2 | 164 | 62 | ||
More Information
Set Membership: |
|

josd2
Genomic Map