Assay Details
| Gene Symbol: fbxo5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044469.1 | NP_001037934.1 | 2 - 3 | 766 | 110 | |
| XM_012963092.1 | XP_012818546.1 | 3 - 4 | 1019 | 110 | ||
| GenBank mRNA | CR761353.2 | - | 2 - 3 | 766 | 110 | |
More Information
Set Membership: |
|

fbxo5
Genomic Map