Assay Details
| Gene Symbol: dnajc24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044478.2 | NP_001037943.2 | 3 - 4 | 301 | 59 | |
| GenBank mRNA | BC168456.1 | - | 3 - 4 | 301 | 59 | |
| CR761411.2 | - | 3 - 4 | 295 | 59 | ||
More Information
Set Membership: |
|

dnajc24
Genomic Map