Assay Details
| Gene Symbol: hes5.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044509.1 | NP_001037974.1 | 1 - 2 | 89 | 65 | |
| GenBank mRNA | BC135288.1 | - | 1 - 2 | 79 | 65 | |
| CR761792.2 | - | 1 - 2 | 89 | 65 | ||
More Information
Set Membership: |
|

hes5.2
Genomic Map