Assay Details
| Gene Symbol: hes5.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044509.1 | NP_001037974.1 | 2 - 3 | 268 | 98 | |
| GenBank mRNA | BC135288.1 | - | 2 - 3 | 258 | 98 | |
| CR761792.2 | - | 2 - 3 | 268 | 98 | ||
More Information
Set Membership: |
|

hes5.2
Genomic Map