Assay Details
| Gene Symbol: ap1s2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045645.2 | NP_001039110.1 | 2 - 3 | 245 | 86 | |
| XM_012957158.2 | XP_012812612.1 | 2 - 3 | 355 | 86 | ||
| XM_012957160.2 | XP_012812614.1 | 2 - 3 | 334 | 86 | ||
| XM_012957161.2 | XP_012812615.1 | 2 - 3 | 341 | 86 | ||
| XM_012957162.2 | XP_012812616.1 | 2 - 3 | 708 | 86 | ||
| GenBank mRNA | BC135368.1 | - | 2 - 3 | 252 | 86 | |
| CR942425.2 | - | 2 - 3 | 247 | 86 | ||
More Information
Set Membership: |
|

ap1s2
Genomic Map