Assay Details
| Gene Symbol: enoph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045647.1 | NP_001039112.1 | 3 - 4 | 650 | 98 | |
| XM_012964336.1 | XP_012819790.1 | 3 - 4 | 538 | 98 | ||
| XM_012964347.2 | XP_012819801.1 | 3 - 4 | 538 | 98 | ||
| XM_012964351.2 | XP_012819805.1 | 2 - 3 | 329 | 98 | ||
| GenBank mRNA | BC118728.1 | - | 3 - 4 | 650 | 98 | |
| CR942427.2 | - | 3 - 4 | 594 | 98 | ||
More Information
Set Membership: |
|

enoph1
Genomic Map