Assay Details
| Gene Symbol: ankrd13c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045661.1 | NP_001039126.1 | 9 - 10 | 1436 | 66 | |
| GenBank mRNA | BC123102.1 | - | 9 - 10 | 1300 | 66 | |
| CR942477.2 | - | 9 - 10 | 1436 | 66 | ||
More Information
Set Membership: |
|

ankrd13c
Genomic Map