Assay Details
| Gene Symbol: rcor1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045668.1 | NP_001039133.1 | 10 - 11 | 1188 | 63 | |
| XM_012968554.2 | XP_012824008.1 | 10 - 11 | 2106 | 63 | ||
| GenBank mRNA | BC160584.1 | - | 10 - 11 | 1154 | 63 | |
| CR942498.2 | - | 10 - 11 | 1188 | 63 | ||
More Information
Set Membership: |
|

rcor1
Genomic Map