Assay Details
| Gene Symbol: ereg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045695.1 | NP_001039160.1 | 2 - 3 | 360 | 78 | |
| XM_012964421.2 | XP_012819875.1 | 2 - 3 | 471 | 78 | ||
| GenBank mRNA | CR942622.2 | - | 2 - 3 | 360 | 78 | |
More Information
Set Membership: |
|

ereg
Genomic Map