Assay Details
| Gene Symbol: arpc5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045732.1 | NP_001039197.1 | 2 - 3 | 560 | 66 | |
| GenBank mRNA | BC122080.1 | - | 2 - 3 | 548 | 66 | |
| CR942333.2 | - | 2 - 3 | 560 | 66 | ||
More Information
Set Membership: |
|

arpc5l
Genomic Map