Assay Details
| Gene Symbol: faf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045770.1 | NP_001039235.1 | 1 - 2 | 84 | 69 | |
| GenBank mRNA | BC157333.1 | - | 1 - 2 | 118 | 69 | |
| CR942713.1 | - | 1 - 2 | 84 | 69 | ||
More Information
Set Membership: |
|

faf2
Genomic Map