Assay Details
| Gene Symbol: atox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045773.3 | NP_001039238.1 | 2 - 3 | 163 | 97 | |
| XM_012959203.2 | XP_012814657.1 | 2 - 3 | 164 | 97 | ||
| GenBank mRNA | BC166972.1 | - | 2 - 3 | 97 | 97 | |
| CN084708.1 | - | 2 - 3 | 166 | 97 | ||
| CR942751.2 | - | 2 - 3 | 92 | 97 | ||
More Information
Set Membership: |
|

atox1
Genomic Map