Assay Details
| Gene Symbol: fam50a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045784.1 | NP_001039249.1 | 1184 | 82 | ||
| GenBank mRNA | BC168785.1 | - | 8 - 9 | 874 | 82 | |
| CR942792.2 | - | 1184 | 82 | |||
More Information
Set Membership: |
|

fam50a
Genomic Map