Assay Details
| Gene Symbol: cat.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078699.1 | NP_001072167.1 | 6 - 7 | 723 | 89 | |
| GenBank mRNA | BC090377.1 | - | 6 - 7 | 714 | 89 | |
| BC123048.1 | - | 6 - 7 | 723 | 89 | ||
More Information
Set Membership: |
|

cat.2
Genomic Map