Assay Details
| Gene Symbol: cat.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078699.1 | NP_001072167.1 | 7 - 8 | 915 | 81 | |
| GenBank mRNA | BC090377.1 | - | 7 - 8 | 906 | 81 | |
| BC123048.1 | - | 7 - 8 | 915 | 81 | ||
More Information
Set Membership: |
|

cat.2
Genomic Map