Assay Details
| Gene Symbol: f8a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078703.1 | NP_001072171.1 | 1 - 2 | 120 | 65 | |
| GenBank mRNA | BC091084.1 | - | 1 - 2 | 50 | 65 | |
| BC122015.1 | - | 1 - 2 | 120 | 65 | ||
More Information
Set Membership: |
|

f8a1
Genomic Map