Assay Details
| Gene Symbol: cwc15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078733.1 | NP_001072201.1 | 6 - 7 | 604 | 70 | |
| XM_012957216.2 | XP_012812670.1 | 6 - 7 | 676 | 70 | ||
| GenBank mRNA | BC118751.1 | - | 6 - 7 | 604 | 70 | |
| BC158969.1 | - | 6 - 7 | 596 | 70 | ||
More Information
Set Membership: |
|

cwc15
Genomic Map