Assay Details
| Gene Symbol: acp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078739.2 | NP_001072207.2 | 2 - 3 | 298 | 56 | |
| GenBank mRNA | BC118766.1 | - | 2 - 3 | 268 | 56 | |
| CX855503.1 | - | 2 - 3 | 298 | 56 | ||
More Information
Set Membership: |
|

acp2
Genomic Map