Assay Details
| Gene Symbol: fn3krp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078748.1 | NP_001072216.1 | 5 - 6 | 623 | 72 | |
| XM_018089717.1 | XP_017945206.1 | 5 - 6 | 521 | 72 | ||
| GenBank mRNA | BC118791.1 | - | 5 - 6 | 623 | 72 | |
More Information
Set Membership: |
|

fn3krp
Genomic Map