Assay Details
| Gene Symbol: c8g.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078777.1 | NP_001072245.1 | 249 | 68 | ||
| GenBank mRNA | BC118854.1 | - | 249 | 68 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

c8g.2| Gene Symbol: c8g.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078777.1 | NP_001072245.1 | 249 | 68 | ||
| GenBank mRNA | BC118854.1 | - | 249 | 68 | ||
Set Membership: |
|