Assay Details
| Gene Symbol: itpkc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078788.1 | NP_001072256.1 | 6 - 7 | 1470 | 66 | |
| GenBank mRNA | BC121240.1 | - | 6 - 7 | 1470 | 66 | |
| CT030585.1 | - | 5 - 6 | 2271 | 66 | ||
More Information
Set Membership: |
|

itpkc
Genomic Map