Assay Details
| Gene Symbol: rnf145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078792.1 | NP_001072260.1 | 9 - 10 | 1642 | 65 | |
| XM_012959210.2 | XP_012814664.1 | 10 - 11 | 1873 | 65 | ||
| XM_012959212.2 | XP_012814666.1 | 10 - 11 | 1856 | 65 | ||
| GenBank mRNA | BC121248.1 | - | 9 - 10 | 1642 | 65 | |
| CT030270.1 | - | 10 - 11 | 1683 | 65 | ||
More Information
Set Membership: |
|

rnf145
Genomic Map