Assay Details
| Gene Symbol: fblim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078804.1 | NP_001072272.1 | 1158 | 97 | ||
| GenBank mRNA | BC121276.1 | - | 1158 | 97 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

fblim1| Gene Symbol: fblim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078804.1 | NP_001072272.1 | 1158 | 97 | ||
| GenBank mRNA | BC121276.1 | - | 1158 | 97 | ||
Set Membership: |
|