Assay Details
| Gene Symbol: ednra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078807.1 | NP_001072275.1 | 3 - 4 | 824 | 56 | |
| GenBank mRNA | BC121283.1 | - | 3 - 4 | 824 | 56 | |
| CU025101.1 | - | 3 - 4 | 719 | 56 | ||
| DQ865481.1 | - | 2 - 3 | 513 | 56 | ||
More Information
Set Membership: |
|

ednra
Genomic Map