Assay Details
| Gene Symbol: phkg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078819.1 | NP_001072287.1 | 4 - 5 | 529 | 66 | |
| XM_012970538.2 | XP_012825992.1 | 4 - 5 | 574 | 66 | ||
| GenBank mRNA | BC121313.1 | - | 4 - 5 | 529 | 66 | |
More Information
Set Membership: |
|

phkg2
Genomic Map