Assay Details
| Gene Symbol: st6galnac4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078839.1 | NP_001072307.1 | 3 - 4 | 367 | 104 | |
| XM_012968605.2 | XP_012824059.1 | 3 - 4 | 451 | 104 | ||
| GenBank mRNA | BC121364.1 | - | 3 - 4 | 367 | 104 | |
More Information
Set Membership: |
|

st6galnac4
Genomic Map