Assay Details
| Gene Symbol: sult4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078854.1 | NP_001072322.1 | 1 - 2 | 365 | 78 | |
| GenBank mRNA | BC121391.1 | - | 1 - 2 | 393 | 78 | |
More Information
Set Membership: |
|

sult4a1
Genomic Map