Assay Details
| Gene Symbol: ccnk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078855.1 | NP_001072323.1 | 10 - 11 | 1198 | 87 | |
| GenBank mRNA | BC121394.1 | - | 10 - 11 | 1198 | 87 | |
More Information
Set Membership: |
|

ccnk
Genomic Map