Assay Details
| Gene Symbol: vsnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078858.1 | NP_001072326.1 | 2 - 3 | 385 | 73 | |
| XM_012970539.2 | XP_012825993.1 | 3 - 4 | 532 | 73 | ||
| GenBank mRNA | BC121397.1 | - | 2 - 3 | 385 | 73 | |
| BC135564.1 | - | 2 - 3 | 325 | 73 | ||
More Information
Set Membership: |
|

vsnl1
Genomic Map