Assay Details
| Gene Symbol: st8sia2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078870.1 | NP_001072338.1 | 4 - 5 | 820 | 81 | |
| GenBank mRNA | BC121420.1 | - | 4 - 5 | 820 | 81 | |
More Information
Set Membership: |
|

st8sia2
Genomic Map