Assay Details
| Gene Symbol: ppp1r16a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078885.1 | NP_001072353.1 | 3 - 4 | 829 | 74 | |
| GenBank mRNA | BC121447.1 | - | 3 - 4 | 829 | 74 | |
More Information
Set Membership: |
|

ppp1r16a
Genomic Map