Assay Details
Gene Symbol: faim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078889.1 | NP_001072357.1 | 6 - 7 | 688 | 65 | |
XM_012957260.2 | XP_012812714.1 | 6 - 7 | 926 | 65 | ||
XM_012957262.2 | XP_012812716.1 | 5 - 6 | 669 | 65 | ||
XM_018091340.1 | XP_017946829.1 | 5 - 6 | 489 | 65 | ||
GenBank mRNA | BC121455.1 | - | 6 - 7 | 688 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
